missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ Easy Polypropylene SPE Column

Brand:  Macherey-Nagel™ 730752

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10670922

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

1 mL
100 mg
80 μm
Solid Phase Extraction Column
650 to 700 m2/g
SPE
Solid Phase Extraction
Chromabond Easy
50 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers