Learn More
Macherey-Nagel™ Chromabond™ CN/SiOH Polypropylene SPE Column
Brand: Macherey-Nagel™ 730112
Code : 50
Additional Details : Weight : 0.50000kg
Specifications
3 mL | |
500 mg | |
Chromabond CN/SiOH | |
p-p Interaction |
SPE | |
Solid Phase Extraction Analysis of PAH | |
Solid Phase Extraction Column |