missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel SPE column CHROMABOND 6mL C6H11ec endcapped and polypropylene column adsorbent weight 1g

Brand:  Macherey Nagel 730444

SPE column CHROMABOND 6mL C6H11ec endcapped and polypropylene column adsorbent weight 1g

Code : 50

Additional Details : Weight : 0.01000kg

Product Code. 10043041

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.