Learn More
CHPG, Tocris Bioscience™
mGlu5 selective agonist
Brand:  Tocris (Bio-Techne) 1049/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13458383
  • £114.75 / 10mg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
A selective mGlu5 metabotropic glutamate receptor agonist, completely inactive at mGlu1a receptors expressed in CHO cells. Active in vivo. CHPG sodium salt (Cat. No. 3695) also available.
Specifications
Chemical Identifiers
170846-74-9
201.606
chpg, 2-chloro-5-hydroxyphenylglycine, rs-2-chloro-5-hydroxyphenylglycine, 2-amino-2-2-chloro-5-hydroxyphenyl acetic acid, chlorohydroxyphenylglycine, d08nqw, r,s-chpg, chpg hplc , white solid, r-2-2-chloro-5-hydroxyphenyl glycine, amino 2-chloro-5-hydroxyphenyl acetic acid
2-amino-2-(2-chloro-5-hydroxyphenyl)acetic acid
C8H8ClNO3
UNIDAFCQFPGYJJ-UHFFFAOYSA-N
3645780
C1=CC(=C(C=C1O)C(C(=O)O)N)Cl
Specifications
CHPG
C8H8ClNO3
UNIDAFCQFPGYJJ-UHFFFAOYSA-N
2-amino-2-(2-chloro-5-hydroxyphenyl)acetic acid
3645780
>99%
170846-74-9
chpg, 2-chloro-5-hydroxyphenylglycine, rs-2-chloro-5-hydroxyphenylglycine, 2-amino-2-2-chloro-5-hydroxyphenyl acetic acid, chlorohydroxyphenylglycine, d08nqw, r,s-chpg, chpg hplc , white solid, r-2-2-chloro-5-hydroxyphenyl glycine, amino 2-chloro-5-hydroxyphenyl acetic acid
C1=CC(=C(C=C1O)C(C(=O)O)N)Cl
201.606
201.61