missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline chloride, 98+% (dry basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 lb.
10 kg
Packaging:
Plastic bottle
Spécification
Melting Point | ∼305°C (decomposition) |
Packaging | Plastic bottle |
Quantity | 5 lb. |
Beilstein | 3563126 |
IUPAC Name | 2-hydroxyethyl(trimethyl)azanium;chloride |
Formula Weight | 139.62 |
Percent Purity | ≥98% |
Physical Form | Crystalline Powder |
Assay | Dry basis |
Chemical Name or Material | Choline chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'