missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline bitartrate, 98+%, Thermo Scientific Chemicals
25.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Description
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.
Solubility
Soluble in alcohol and slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Choline Bitartrate is used by nerve cells to manufacture a chemical called acetylcholine, which effectively allows cells to communicate with each other. Choline is also heavily associated with neurological function and can help improve movement, coordination, and myofibril contraction.
Solubility
Soluble in alcohol and slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 151°C to 153°C |
Color | White |
Quantity | 250 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in alcohol and slightly soluble in water. |
Formula Weight | 253.25 |
Percent Purity | ≥98% |
Physical Form | Powder |
Chemical Name or Material | Choline bitartrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'