missing translation for 'onlineSavingsMsg'
Learn More

Cholic Acid, Sodium Salt, 99%, ACROS Organics™

Brand:  Acros Organics 405195000

Code : 29

Additional Details : CAS Number : 361-09-1 Weight : 0.55000kg

Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10667954

  • £238.50 / 500g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Cholic acid, sodium salt
98.0 to 102.0% (on dried basis)
C24H39NaO5
MFCD00064138
361-09-1
99%
430.55
3α, 7α, 12α-Trihydroxy-5β-cholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid, sodium salt

Specifications

Cholic acid, sodium salt
361-09-1
5.0% max. (110°C)
Glass bottle
Powder
99%
430.55
+ 35.00 (20.00°C c=0.6,ethanol)
White to off-white
3α, 7α, 12α-Trihydroxy-5β-cholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid, sodium salt
sodium deoxycholate: < 2.0%
50ppm max.
Authentic
+33° to +37° (20°C, 589nm) (c=0.6, ethanol)
98.0 to 102.0% (on dried basis)
C24H39NaO5
MFCD00064138
14, 2203
7.5 to 9.5 (5% aq. soln.)
500g
Videos
SDS