missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cholesteryl oleate

Brand:  Alfa Aesar™ A11378.14

Code : NEW

Additional Details : CAS Number : 303-43-5 Weight : 0.01000kg

Quantity 25g

 View more versions of this product

Product Code. 11480193

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Applications
Cholesteryl oleate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is also used in chemical research.

Solubility
It is soluble in chloroform: 0.1 g/Ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Incompatible to strong oxidizing agents.
Specifications

Chemical Identifiers

Cholesteryl oleate
C45H78O2
MFCD00003645
Oleic acid cholesteryl ester
303-43-5
651.12
2343071

Specifications

Cholesteryl oleate
C45H78O2
MFCD00003645
Oleic acid cholesteryl ester
25g
−24° (c=1 in chloroform)
303-43-5
651.12
2343071
44° to 47°C
Odorless
Videos
SDS