missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7080759
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
910-31-6
C27H45Cl
405.107
MFCD00003632
OTVRYZXVVMZHHW-DGPGEHMFSA-N
cholesterylchloride
131632519
(8S,9R,10R,13R,14S,17R)-3-chloro-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)Cl)C)C
This item is not returnable. View return policy
910-31-6
C27H45Cl
405.107
MFCD00003632
OTVRYZXVVMZHHW-DGPGEHMFSA-N
cholesterylchloride
131632519
(8S,9R,10R,13R,14S,17R)-3-chloro-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene
CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)Cl)C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 94°C to 96°C
Quantity 25 g
Beilstein 2703655
Optical Rotation −26° (c=1 in benzene)
Formula Weight 405.11
Percent Purity 98%
Chemical Name or Material Cholesteryl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'