missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol, 95%, ACROS Organics™

Brand:  Acros Organics 110195000

Code : 29

Additional Details : CAS Number : 57-88-5 Weight : 0.55000kg

Packaging Plastic bottle
Quantity 500 g

 View more versions of this product

Product Code. 10080790

  • £164.00 / 500g

In stock, ready to ship: 9

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

57-88-5
386.66
cholestrin, cholest-5-en-3beta-ol, provitamin d, dusoline, cholesterin, dusoran, cholesteryl alcohol, cordulan, cholesterol, cholesterine
CHEBI:16113
CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
C27H46O
HVYWMOMLDIMFJA-DPAQBDIFSA-N
5997
(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol

Specifications

Cholesterol
Plastic bottle
500 g
57-88-5
C27H46O
15, 2208
HVYWMOMLDIMFJA-DPAQBDIFSA-N
(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
5997
386.65
1.06 g/mL
-36.00 (20.00°C=2,dioxane)
Stabilized
95%
01,141
cholestrin, cholest-5-en-3beta-ol, provitamin d, dusoline, cholesterin, dusoran, cholesteryl alcohol, cordulan, cholesterol, cholesterine
CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
386.66
CHEBI:16113
Videos
SDS Documents