missing translation for 'onlineSavingsMsg'
Learn More

Chlortetracycline hydrochloride, conforms to Eur. Ph., ACROS Organics™

Brand:  Acros Organics 430960050

Code : 29

Additional Details : CAS Number : 64-72-2 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10882041

  • £20.50 / 5g


EU Stock 37
Add to basket


Specifications

Chemical Identifiers

Chlortetracycline hydrochloride
C22H23ClN2O8·HCl
MFCD00082440
94.5 to 102.0% (sum of the contents of chlortetra cycline hydrochloride and tetracycline hydrochloride, dry substance), ≥89.5% (chlortetracycline hydrochloride, dry substance), 8% max. (tetracycline hydrochloride, dry substance)
515.33
CTC

Specifications

0.40 max. (460nm)
− 242.50 (20.00°C c=0.5,water)
220.0°C
2.3 to 3.3 (1%, water CO2 free)
50ppm max.
Passes Test
0.5% max.
2% Max.
94.5 to 102.0% (sum of the contents of chlortetra cycline hydrochloride and tetracycline hydrochloride, dry substance), ≥89.5% (chlortetracycline hydrochloride, dry substance), 8% max. (tetracycline hydrochloride, dry substance)
515.33
CTC
Chlortetracycline hydrochloride
Yellow
5g
64-72-2
Authentic
Glass bottle
− 242.50
Powder
C22H23ClN2O8·HCl
MFCD00082440
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Chlortetracycline hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS