missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotris(triphenylphosphine)rhodium(I), 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Chlorotris(triphenylphosphine)rhodium(I) is used as a homogeneous hydrogenation catalyst. It is used in the selective hydrogenation of alkenes and alkynes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∼250°C (decomposition) |
Color | Red |
Odor | Odorless |
Linear Formula | RhCl[(C6H5)3P]3 |
Quantity | 1 g |
Merck Index | 14,10047 |
Solubility Information | Soluble in most solvents (e;g; benzene,ethanol,chloroform,dichloromethane) but with phosphine dissociation; Reacts with O2 in solution |
Formula Weight | 925.23 |
Percent Purity | 97% |
Physical Form | Micro Crystals |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'