missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriethylsilane, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022664
Click to view available options
Quantity:
10 g
50 g
250 g
994-30-9
C6H15ClSi
150.72
MFCD00000507
DCFKHNIGBAHNSS-UHFFFAOYSA-N
triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
13819
chloro(triethyl)silane
CC[Si](Cl)(CC)CC
994-30-9
C6H15ClSi
150.72
MFCD00000507
DCFKHNIGBAHNSS-UHFFFAOYSA-N
triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
13819
chloro(triethyl)silane
CC[Si](Cl)(CC)CC

Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

Solubility
Soluble in aprotic organic solvents.

Notes
Incompatible with bases, strong oxidizing agents, strong acids, alcohols, amines, ammonia, aldehydes and acetone.
TRUSTED_SUSTAINABILITY
Boiling Point 145°C to 147°C
Melting Point ∼-75°C
Quantity 10 g
Linear Formula (CH3CH2)3SiCl
UN Number UN2986
Beilstein 1732860
Flash Point 29°C (84°F)
Solubility Information Soluble in aprotic organic solvents.
Formula Weight 150.73
Percent Purity ≥98%
Odor Pungent
Refractive Index 1.431
Sensitivity Moisture sensitive
Density 0.899
Chemical Name or Material Chlorotriethylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'