missing translation for 'onlineSavingsMsg'
Learn More

Chlorosulfonic acid, typically 99%, Alfa Aesar™

POA - £192.00

Chemical Identifiers

CAS 7790-94-5
Molecular Formula ClHO3S
Molecular Weight (g/mol) 116.515
MDL Number MFCD00011523
InChI Key XTHPWXDJESJLNJ-UHFFFAOYSA-N
Synonym chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909
PubChem CID 24638
IUPAC Name sulfurochloridic acid
SMILES OS(=O)(=O)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492977
SDS View Documents
Alfa Aesar
087977.A1
1kg
£192.00
1kg
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
15402987
SDS View Documents
Alfa Aesar
087977.D5
3 x 1kg
POA
Each
View alternative products
Save Up to 
»
N/A Please to purchase this item. Need a web account? Register with us today!
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
15482977
SDS View Documents
Alfa Aesar
087977.30
250g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

7790-94-5
116.515
XTHPWXDJESJLNJ-UHFFFAOYSA-N
24638
OS(=O)(=O)Cl
ClHO3S
MFCD00011523
chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909
sulfurochloridic acid

Specifications

Chlorosulfonic Acid
Yellow to Brown
7790-94-5
typically 99%
MFCD00011523
chlorosulfonic acid, chlorosulfuric acid, sulfuric chlorohydrin, chlorosulphuric acid, chlorosulphonic acid, monochlorosulfuric acid, unii-2o9axl1tj4, sulfonic acid, monochloride, chloridosulfuric acid, hsdb 909
XTHPWXDJESJLNJ-UHFFFAOYSA-N
sulfurochloridic acid
116.515
116.52
Odorless
-80°C
1.433
1kg
Yellow to brown liquid
ClHO3S
1754
Miscible with hydrocarbons.
OS(=O)(=O)Cl
1.753 g/cm3 at 20°C
24638
Liquid
Moisture sensitive
158°C
SDS
Product Certifications