missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorosulfonic acid, typically 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
3 x 1 kg
Description
Chlorosulfonic acid is used as detergent and as an anti-contrail agent. It is used as an intermediate in the production of other substances such as pharmaceuticals and chemicals. It finds application in producing smoke screens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Appearance | Yellow to brown liquid |
Color | Yellow to Brown |
Boiling Point | 158°C |
Density | 1.753 |
Odor | Odorless |
Assay Percent Range | typically 99% |
Quantity | 3 x 1 kg |
Physical Form | Liquid |
UN Number | 1754 |
Solubility Information | Miscible with hydrocarbons. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'