missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroplatinic Acid Hexahydrate, ACS Reagent, ≥37.50% Pt basis, Honeywell™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 25 g |
| Linear Formula | H2PtCl6 · 6H2O |
| UN Number | UN2507 |
| Formula Weight | 517.9g/mol |
| Percent Purity | ≥37.50% (Pt basis) |
| Grade | ACS Reagent |
| Packaging | Glass Bottle |
| Chemical Name or Material | Chloroplatinic Acid Hexahydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction