Alfa Aesar™ Chlorophenol Red sodium salt, 0.04% w/v aq. soln.


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 038699.AE

CHLOROPHENOL RED SODIUM .04% AQUEOUS SOL. 100ML

UNSPSC: 12352005

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg


Quantity 100mL

Product Code. 11348967

 N/A
N/A

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description and Specification

Chemical Identifiers

Chemical Name or Material Chlorophenol Red sodium salt
Formula Weight 445.26
MDL Number MFCD00151199

Specification

Formula Weight 445.26
Chemical Name or Material Chlorophenol Red sodium salt
Quantity 100mL
TSCA TSCA
Concentration 0.04% w/v aq. soln.
Physical Form Liquid
MDL Number MFCD00151199
Density 0.990g/mL