missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl pivalate, 99+%, Thermo Scientific Chemicals

Catalog Number p-3757243
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18997-19-8
C6H11ClO2
150.60
MFCD00000884
GGRHYQCXXYLUTL-UHFFFAOYSA-N
chloromethyl pivalate, propanoic acid, 2,2-dimethyl-, chloromethyl ester, pivalic acid chloromethyl ester, pivaloyloxymethyl chloride, pivaloylmethyl chloride, chloromethyl trimethylacetate, 2,2-dimethylpropionic acid chloromethyl ester, chioromethyl pivalate, chloromethyl pivaloate, chloro-methyl pivalate
87885
chloromethyl 2,2-dimethylpropanoate
CC(C)(C)C(=O)OCCl
This item is not returnable. View return policy
18997-19-8
C6H11ClO2
150.60
MFCD00000884
GGRHYQCXXYLUTL-UHFFFAOYSA-N
chloromethyl pivalate, propanoic acid, 2,2-dimethyl-, chloromethyl ester, pivalic acid chloromethyl ester, pivaloyloxymethyl chloride, pivaloylmethyl chloride, chloromethyl trimethylacetate, 2,2-dimethylpropionic acid chloromethyl ester, chioromethyl pivalate, chloromethyl pivaloate, chloro-methyl pivalate
87885
chloromethyl 2,2-dimethylpropanoate
CC(C)(C)C(=O)OCCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0380g/mL
Boiling Point 146°C to 148°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCO2CH2Cl
Refractive Index 1.4160 to 1.418
Quantity 100 g
Specific Gravity 1.038
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,toluene and chlorinated,hydrocarbons
Formula Weight 150.6
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Chloromethyl pivalate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloromethyl pivalate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'