missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)triphenylphosphonium chloride, 95%, tech., Thermo Scientific Chemicals

Catalog Number p-88326
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5293-84-5
C19H17Cl2P
347.22
MFCD00011867
SXYFAZGVNNYGJQ-UHFFFAOYSA-M
chloromethyl triphenylphosphonium chloride, chloromethyltriphenylphosphonium chloride, chloromethyl triphenylphosphoniumchloride, phosphonium, chloromethyltriphenyl-, chloride, chloromethly triphenylphosphonium chloride, chloromethyl triphenylphosphanium chloride, chloromethyl triphenylphosphonium chliride, triphenyl chloromethyl-phosphonium chloride, acmc-1albk, phosphonium, chloromethyl triphenyl-, chloride
197823
chloromethyl(triphenyl)phosphanium;chloride
[Cl-].ClC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5293-84-5
C19H17Cl2P
347.22
MFCD00011867
SXYFAZGVNNYGJQ-UHFFFAOYSA-M
chloromethyl triphenylphosphonium chloride, chloromethyltriphenylphosphonium chloride, chloromethyl triphenylphosphoniumchloride, phosphonium, chloromethyltriphenyl-, chloride, chloromethly triphenylphosphonium chloride, chloromethyl triphenylphosphanium chloride, chloromethyl triphenylphosphonium chliride, triphenyl chloromethyl-phosphonium chloride, acmc-1albk, phosphonium, chloromethyl triphenyl-, chloride
197823
chloromethyl(triphenyl)phosphanium;chloride
[Cl-].ClC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 258.0°C to 264.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula ClCH2P(C6H5)3Cl
Quantity 5 g
Fieser 07,62
Formula Weight 347.21
Percent Purity 95%
Grade Technical
Physical Form Fine Crystalline Powder
Chemical Name or Material (Chloromethyl)triphenylphosphonium chloride, tech., 95%
Show More Show Less
compliance-icons
Product Identifier
  • (Chloromethyl)triphenylphosphonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'