missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ReagentPlus™, ≥99.8%, Honeywell™

Catalog Number 15601290
Click to view available options
Quantity:
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
67-66-3
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl
This item is not returnable. View return policy
67-66-3
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl

Reagent Grade, contains 0.5-1.0% ethanol as stabilizer,≥99.8%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -63°C
UN Number UN1888
Beilstein 1731042
Formula Weight 119.38g/mol
Percent Purity ≥99.8%
Grade ReagentPlus™
Density 1.492 g/mL at 25°C, 1.48 g/mL at 25°C
Boiling Point 60°C to 62°C
Quantity 10 L
Packaging Stainless Steel Can
Physical Form Liquid
Refractive Index n20/D 1.445(lit.)
Vapor Pressure 160 mmHg (20°C)
Chemical Name or Material Chloroform
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'