missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Reagent ACS, Spectro Grade, 99.8%, Thermo Scientific Chemicals
24.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
Specifications
Melting Point | -63.0°C |
Absorbance | 0.01 max. at 290nm, 0.01 max. at 400nm, 0.05 max. at 270nm, 0.15 max. at 260nm, 0.25 max. at 255nm, 1 max. at 245nm |
Decomposition Information | 290°C |
Density | 1.4800g/mL |
Boiling Point | 61.0°C |
Quantity | 500 mL |
Packaging | EcoSafPak™ Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.4443 to 1.4463 |
Assay Percent Range | 99.8% |
Show More |
Safety and Handling
Product Identifier
- Chloroform, stabilized with ethanol
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : Chloroform
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'