missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, Hydrocarbone Stabilized, BAKER ANALYZED HPLC, J.T. Baker™
135.85 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.
- Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
- Low backgrounds free of extraneous peaks
- Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
- Fluorescence testing for trace impurities which would cause interference
- Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
- Lot to lot consistency, with an actual Lot Analysis on the label
- Innovative packaging options to assure solvent quality to the point-of-use
Specifications
Melting Point | -63°C |
Absorbance | Max. 0.15 at 254nm, Max. 0.05 at 280nm, Max. 0.01 at 350nm |
Color | Colorless |
Solubility Information | 8g/L in water |
IUPAC Name | trichloromethane |
Formula Weight | 119.38g/mol |
Percent Purity | ≥99.8% |
Grade | BAKER ANALYZED™ HPLC |
Density | 1.483g/mL at 20°C |
Boiling Point | 62°C |
Show More |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P281-Use personal protective equipment as required.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'