missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, HPLC Grade, 99.5+% min, stab. with amylene, Thermo Scientific Chemicals

Catalog Number 11300278
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
This item is not returnable. View return policy
67-66-3
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
ClC(Cl)Cl
This item is not returnable. View return policy
67-66-3
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
ClC(Cl)Cl

Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 63°C
Color Colorless
Density 1.492
Boiling Point 60.5°C to 61.5°C
Quantity 4 L
Ignition Point 982°C (1800°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.446
Assay Percent Range >99.5%
Vapor Pressure 158mm Hg at 20°C
Beilstein 1731042
Solubility Information Miscible with diethyl ether,oils,ligroin,alcohol,carbon tetrachloride and carbon disulfide.
Formula Weight 119.38
Grade HPLC
Chemical Name or Material Chloroform
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'