missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene, Fisher Chemical™

Catalog Number p-7072921
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
67-66-3
CHCl3
119.37
826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
ClC(Cl)Cl
This item is not returnable. View return policy
67-66-3
CHCl3
119.37
826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
ClC(Cl)Cl

Chloroform, Extra Dry, for Synthesis, Stabilised with Amylene

TRUSTED_SUSTAINABILITY
Melting Point -63°C
Color Colorless
Boiling Point 61°C
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 213mbar at 20°C
Viscosity 0.56 mPaS at 20°C
Formula Weight 119.38g/mol
compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with amylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'