missing translation for 'onlineSavingsMsg'
Learn More

Chloroform-d, 100% (Isotopic), contains 0.03% v/v TMS, Thermo Scientific Chemicals

Catalog Number p-7051509
33.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 Ea.
5 Ea.
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
865-49-6
CHCl3
120.375
MFCD00000827
HEDRZPFGACZZDS-MICDWDOJSA-N
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
71583
CHEBI:85365
trichloro(deuterio)methane
C(Cl)(Cl)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
865-49-6
CHCl3
120.375
MFCD00000827
HEDRZPFGACZZDS-MICDWDOJSA-N
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
71583
CHEBI:85365
trichloro(deuterio)methane
C(Cl)(Cl)Cl

Widely used in the organic solvent for the NMR analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 1.50 g/mL
Refractive Index 1.4445
Sensitivity Light and Moisture
Quantity 5 Ea.
Melting Point -64°C
Boiling Point 62°C
UN Number UN1888
Solubility Information Miscible with organic solvents ethyl acetate,acetone.
Formula Weight 120.38
Percent Purity 100%
Concentration or Composition (by Analyte or Components) contains 0.03% v/v TMS
Chemical Name or Material Chloroform-d, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'