missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Riedel-de Haen™ Chloroform, CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer, ≥99.8%, Honeywell Riedel-de Haën™

CHROMASOLV™ for residue analysis, contains ~1% ethanol as stabilizer,≥99.8%

£48.09 - £351.99

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.369
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES C(Cl)(Cl)Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15652770 SDS View Documents Honeywell Riedel-de Haen™
25669-1L
1L Glass Bottle £48.09
1 L View alternative products
Save Up to 
»
EU Stock 4 Please to purchase this item. Need a web account? Register with us today!
15662770 SDS View Documents Honeywell Riedel-de Haen™
25669-2.5L
2.5L Glass Bottle £101.05
2.50 L View alternative products
Save Up to 
»
EU Stock 96 Please to purchase this item. Need a web account? Register with us today!
15672770 SDS View Documents Honeywell Riedel-de Haen™
25669-4X2.5L
4 x 2.5L Glass Bottle This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
15682770 SDS View Documents Honeywell Riedel-de Haen™
25669-6X1L
6 x 1L Glass Bottle This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Specifications

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
C(Cl)(Cl)Cl

Specifications

Chloroform
64-17-5
<1.0000%
CHCl3
MFCD00000826
1731042
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
>99.8%
1.492 g/mL at 25 °C(lit.)
60°C to 62°C
1L
CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer, ≥99.8%
≥0.1000%
GC product+ethanol
CHCl3
UN1888
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
C(Cl)(Cl)Cl
119.369
CHEBI:35255
Residue Analysis
Glass Bottle
-63°C (literature)