missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer, ≥99.8%, Honeywell Riedel-de Haën™

Catalog Number 15672770
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
67-66-3, 64-17-5
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl
This item is not returnable. View return policy
67-66-3, 64-17-5
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl

CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer,≥99.8%

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY
Melting Point -63°C (literature)
CAS Min % ≥90.0000%, ≥0.1000%
CAS Max % ≤100.0000%, <1.0000%
Linear Formula CHCl3
UN Number UN1888
Beilstein 1731042
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
Percent Purity >99.8%
Grade Residue Analysis
Density 1.492 g/mL at 25 °C(lit.)
Boiling Point 60°C to 62°C
Quantity 4 x 2.5 L
Packaging Glass Bottle
Assay GC product+ethanol
Chemical Name or Material Chloroform, CHROMASOLV™ for residue analysis, contains ∼1% ethanol as stabilizer, ≥99.8%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'