missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, BAKER ANALYZED™, J.T. Baker™
163.40 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Melting Point | -63°C |
Color | Max. 10 (APHA) |
Assay Percent Range | min. 99% |
Density | 1.4832 g/cm³ (20°C) |
Boiling Point | 61.7°C (1013 hPa) |
Quantity | 1 L |
Packaging | Glass Bottle |
Residue after Evaporation | max. 0.001% |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P281-Use personal protective equipment as required.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'