Chloroform 99%, ACROS Organics™


Overview
Brand: GC Internal (Acros/Geel)

Manufacturer Part Number: 158210100

UNSPSC: 12190000

Code: 50

Additional Details:
Additional Details: CAS Number: 67-66-3 Weight: 14.90000kg Transport: UN number : 1888 Chem class : 1 Pack group : III


Packaging Plastic Jerrycan
Quantity 10L

Product Code. 10139172

Quantity Price
1 £ 104.7 / 10 L
Estimated Shipment
28-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Chloroform
CAS 67-66-3
CAS 64-17-5
Purity Grade Extra Pure
Assay ≥99.0% (excluding stabilizer)
Assay Percent Range 99+%
Molecular Formula CHCl3
Linear Formula CHCl3
Formula Weight 119.38
MDL Number MFCD00000826

Specification

Assay ≥99.0% (excluding stabilizer)
Formula Weight 119.38
Purity Grade Extra Pure
Physical Form Liquid
Quantity 10L
Color Clear
Beilstein 01, 61
Merck Index 15, 2142
Packaging Plastic Jerrycan
Linear Formula CHCl3
Residue after Evaporation 0.002% max.
Health Hazard 3 GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Additional Information Vapor Pressure: 213mbar at 20°C
Decomposition Information 290°C
CAS 67-66-3
Assay Percent Range 99+%
MDL Number MFCD00000826
Stabilizer Approx. 1% ethanol
Health Hazard 2 GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
Water 0.02% max. (K.F.)
Fieser 01,130; 08,92; 15,84
Molecular Formula CHCl3
Health Hazard 1 Warning
Density 1.4800g/mL
Chemical Name or Material Chloroform
CAS 64-17-5
Documentation Safety and Handling

Warning

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.