missing translation for 'onlineSavingsMsg'
Learn More

chlorodiphenylphosphine, 98%, Thermo Scientific Chemicals

Catalog Number p-3735968
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1079-66-9
C12H10ClP
220.64
MFCD00000529
XGRJZXREYAXTGV-UHFFFAOYSA-N
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
66180
chloro(diphenyl)phosphane
ClP(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1079-66-9
C12H10ClP
220.64
MFCD00000529
XGRJZXREYAXTGV-UHFFFAOYSA-N
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
66180
chloro(diphenyl)phosphane
ClP(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2290g/mL
Boiling Point 320.0°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2PCl
Refractive Index 1.6340 to 1.6370
Quantity 5 g
Beilstein 16, 763
Fieser 11,120; 16,77
Merck Index 15, 2139
Specific Gravity 1.229
Solubility Information Solubility in water: reacts violently with water
Formula Weight 220.64
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chlorodiphenylphosphine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chlorodiphenylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'