missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylphosphine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7051495
18.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1079-66-9
C12H10ClP
220.64
MFCD00000529
XGRJZXREYAXTGV-UHFFFAOYSA-N
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
66180
chloro(diphenyl)phosphane
ClP(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1079-66-9
C12H10ClP
220.64
MFCD00000529
XGRJZXREYAXTGV-UHFFFAOYSA-N
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
66180
chloro(diphenyl)phosphane
ClP(C1=CC=CC=C1)C1=CC=CC=C1

Chlorodiphenylphosphine is used in the synthesis of various phosphines and an useful reagent in for the introduction of Ph2P group. It is also useful as an intermediate to make antioxidants, flame retardants, stabilizers, catalysts, photoinitiators and optical brighteners. It is also used to produce diphenylphosphinous acid methyl esters. It is used as a precursor in the preparation of various phosphines. It is involved in the preparation of phosphines, which is used as stabilizers for plastics, flame retardants and UV-hardening paint systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 14°C to 16°C
Color Colorless to Yellow
Density 1.204 g/mL
Boiling Point 318°C to 321°C
Flash Point 138°C (280°F)
Odor Acrid
Linear Formula (C6H5)2PCl
Refractive Index 1.636
Quantity 10 g
UN Number UN3265
Beilstein 512032
Sensitivity Air and moisture sensitive
Solubility Information Miscible with alcohol. Slightly miscible with ammonia.
Formula Weight 220.64
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Chlorodiphenylphosphine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'