missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylphosphine, 95%, tech., ACROS Organics™

£40.08 - £325.00

Chemical Identifiers

CAS 1079-66-9
Molecular Formula C12H10ClP
Molecular Weight (g/mol) 220.64
MDL Number MFCD00000529
InChI Key XGRJZXREYAXTGV-UHFFFAOYSA-N
Synonym chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
PubChem CID 66180
IUPAC Name chloro(diphenyl)phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10071020
SDS View Documents
Acros Organics
109130250
25mL Glass bottle
£40.08
25mL
View alternative products
Save Up to 
»
In stock, ready to ship: 4 Please to purchase this item. Need a web account? Register with us today!
10081020
SDS View Documents
Acros Organics
109131000
100mL Glass bottle
£89.51
100mL
View alternative products
Save Up to 
»
In stock, ready to ship: 7 Please to purchase this item. Need a web account? Register with us today!
10637364
SDS View Documents
Acros Organics
109135000
500mL Glass bottle
£325.00
500mL
View alternative products
Save Up to 
»
Estimated Shipment
08-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C-X bond formation reagent
Specifications

Chemical Identifiers

1079-66-9
220.64
XGRJZXREYAXTGV-UHFFFAOYSA-N
66180
C1=CC=C(C=C1)P(C2=CC=CC=C2)Cl
C12H10ClP
MFCD00000529
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
chloro(diphenyl)phosphane

Specifications

Chlorodiphenylphosphine
138°C
1.6340 to 1.6370
320.0°C
1079-66-9
C12H10ClP
MFCD00000529
11,120; 16,77
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
XGRJZXREYAXTGV-UHFFFAOYSA-N
chloro(diphenyl)phosphane
66180
95%
1.2290g/mL
Glass bottle
1.229
25mL
94% min. (GC)
(C6H5)2PCl
16, 763
15, 2139
Solubility in water: reacts violently with water
C1=CC=C(C=C1)P(C2=CC=CC=C2)Cl
220.64
220.64
Technical
Safety and Handling

Safety and Handling

Product Identifier
  • Chlorodiphenylphosphine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS