missing translation for 'onlineSavingsMsg'
Learn More

chlorodiphenylmethane, 98%, Thermo Scientific Chemicals

Catalog Number 10478500
Click to view available options
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
90-99-3
C13H11Cl
202.68
MFCD00000855
ZDVDCDLBOLSVGM-UHFFFAOYSA-N
benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene
7035
[chloro(phenyl)methyl]benzene
C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl
This item is not returnable. View return policy
90-99-3
C13H11Cl
202.68
MFCD00000855
ZDVDCDLBOLSVGM-UHFFFAOYSA-N
benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene
7035
[chloro(phenyl)methyl]benzene
C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 15°C to 17°C
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 270°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2CHCl
Refractive Index 1.5940 to 1.597
Quantity 5 mL
Specific Gravity 1.14
Solubility Information Solubility in water: slightly soluble
Formula Weight 202.68
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chlorodiphenylmethane
Show More Show Less
compliance-icons
Product Identifier
  • Chlorodiphenylmethane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'