chlorodimethylthexylsilane, 95%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 408600050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 67373-56-2 Weight: 0.06000kg Transport: UN number : 2920 Chem class : 1a Pack group : II


Packaging Glass bottle
Quantity 5g

Product Code. 10157470

Quantity Price
1 £ 16.1 / 5g
EU Stock 3
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material chlorodimethylthexylsilane
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H19ClSi
Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl
Formula Weight 178.78
MDL Number MFCD00012175
Synonym Dimethylthexylsilyl chloride

Specification

Assay ≥94%
Formula Weight 178.78
Physical Form Liquid
Quantity 5g
Color Clear colorless to almost colorless
Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 67373-56-2
Refractive Index 1.4480 to 1.4500 (20°C, 589nm)
MDL Number MFCD00012175
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Packaging Glass bottle
Molecular Formula C8H19ClSi
Health Hazard 1 GHS Signal Word: Danger
Density 0.9g/mL
Synonym Dimethylthexylsilyl chloride
Chemical Name or Material chlorodimethylthexylsilane
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.