missing translation for 'onlineSavingsMsg'
Learn More

chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium, 97%, ACROS Organics™

Brand:  Acros Organics 435120010

Code : 29

Additional Details : CAS Number : 791629-96-4 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 11947041

  • £262.30 / 1g


Estimated Shipment
07-02-2018
Add to basket


Specifications

Chemical Identifiers

chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium
97%
C16H37NP2·ClH2Ir
535.1

Specifications

chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium
240.6° to 241.9°C
791629-96-4
Authentic
Powder
97%
535.1
White to tan
1g
34.50 to 37.30% C, 2.50 to 2.70% N
Glass bottle
97%
C16H37NP2·ClH2Ir
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Chlorodihydrido(bis((2-diisopropylphosphino)ethyl)amine)iridium(III)

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS