missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(cyclooctene)iridium(I) dimer, Ir nominally 42.9%, Thermo Scientific Chemicals

Catalog Number p-7059598
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
12246-51-4
C32H56Cl2Ir2-2
896.134
MFCD00213465
CJJIQMGSHWWMCK-XFCUKONHSA-L
chlorobis cyclooctene iridium i dimer, di-mu-chlorotetrakis cyclooctene diiridium i, bis cyclooctene iridium i chloride, dimer, chlorobis cyclooctene-iridium i dimer, chlorobis cyclooctene iridium dimer, di-mu-chlorobis cyclooctene iridium i
91972094
cyclooctene;iridium;dichloride
C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.[Cl-].[Cl-].[Ir].[Ir]
This item is not returnable. View return policy
12246-51-4
C32H56Cl2Ir2-2
896.134
MFCD00213465
CJJIQMGSHWWMCK-XFCUKONHSA-L
chlorobis cyclooctene iridium i dimer, di-mu-chlorotetrakis cyclooctene diiridium i, bis cyclooctene iridium i chloride, dimer, chlorobis cyclooctene-iridium i dimer, chlorobis cyclooctene iridium dimer, di-mu-chlorobis cyclooctene iridium i
91972094
cyclooctene;iridium;dichloride
C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.[Cl-].[Cl-].[Ir].[Ir]

Chlorobis(cyclooctene)iridium(I) dime is used as catalyst for Isomerization-hydroboration reactions with nido-carboranyldiphosphine as stabilizing ligand, Hydrogen peroxide oxidation of hydroxamic acids and their subsequent hetero Diels-Alder cycloaddition reactions, Immobilization of organic functional groups onto solid supports through vinylsilane coupling reactions, Alkylation reactions, Guerbet reaction, Allylic amination reactions in a DNA-diene-iridium(I) hybrid system, Asymmetric hydroamination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Soluble in organic solvents.
Formula Weight 896
Concentration or Composition (by Analyte or Components) Ir nominally 42.9%
Chemical Name or Material Chlorobis (cyclooctene)iridium (I) dimer

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'