missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetonitrile, 98+%, Alfa Aesar™

Brand:  Alfa Aesar A12825.A1

Code : NEW

Additional Details : CAS Number : 107-14-2 Weight : 1.00000kg Transport : UN number : 2668 Chem class : 6.1 Pack group : I

Quantity 1kg

 View more versions of this product

Product Code. 15403067

  • £252.00 / 1kg

Estimated Shipment: 10-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
Chloroacetonitrile is used in the electrochemical synthesis of cyanoacetic acid with carbon dioxide. It is involved in phase-transfer-catalyzed Darzen′s condensation reaction with cyclohexanone. It is also used as an eluent additive in thermospray liquid chromatography/mass spectrometry. Further, it is used to prepare polysubstituted pyrido[1,2-a]benzimidazole by reacting with other reactant such as malononitrile, aromatic aldehyde and pyridine.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and acids.
Specifications

Chemical Identifiers

107-14-2
75.495
RENMDAKOXSCIGH-UHFFFAOYSA-N
7856
2-chloroacetonitrile
C2H2ClN
MFCD00001885
chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro
CHEBI:82416
C(C#N)Cl

Specifications

Chloroacetonitrile
56°C (132°F)
124°C to 126°C
107-14-2
ClCH2CN
UN2668
chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro
RENMDAKOXSCIGH-UHFFFAOYSA-N
2-chloroacetonitrile
7856
75.5
1.195
1.423
1kg
C2H2ClN
MFCD00001885
506028
Immiscible with water.
C(C#N)Cl
75.495
CHEBI:82416
≥98%
Videos
SDS Documents
Certificates