missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetone, 96%, stabilized, ACROS Organics™

Brand:  Acros Organics 173470050

Code : 29

Additional Details : CAS Number : 78-95-5 Weight : 5.85000kg Transport : UN number : 1695 Chem class : 6.1 Pack group : I

Quantity 5L
Packaging Plastic drum

 View more versions of this product

Product Code. 10697953

  • £281.50 / 5 L


EU Stock 1
Add to basket

Sea freight only: For EU stock allow up to 15 days delivery.

Specifications

Chemical Identifiers

Chloroacetone
78-95-5
96%
ClCH2COCH3
MFCD00000936
Stabilized
96%
C3H5ClO
92.52
Chloro-2-propanone

Specifications

Chloroacetone
32°C
119.0°C
− 44.5°C
Stabilized
Authentic
1.4310 to 1.4330
Solubility in water: 124g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
96%
C3H5ClO
92.52
01, 653
Chloro-2-propanone
1.1600g/mL
1.16
Clear yellow to yellow-brown
5L
78-95-5
Plastic drum
Liquid
96%
0.1% epoxidized soybean oil
ClCH2COCH3
MFCD00000936
15, 2114
Videos
SDS