missing translation for 'onlineSavingsMsg'
Learn More

chloroacetic anhydride, 97%, Thermo Scientific Chemicals

Catalog Number 10474115
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
541-88-8
C4H4Cl2O3
170.97
MFCD00000929
PNVPNXKRAUBJGW-UHFFFAOYSA-N
chloroacetic anhydride, chloracetic anhydride, monochloroacetic acid anhydride, 2-chloroacetic anhydride, chloroacetic acid anhydride, chloroacetyl anhydride, acetic acid, chloro-, anhydride, unii-8q7txh7r5f, monochloroacetic anhydride, 8q7txh7r5f
10946
(2-chloroacetyl) 2-chloroacetate
ClCC(=O)OC(=O)CCl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
541-88-8
C4H4Cl2O3
170.97
MFCD00000929
PNVPNXKRAUBJGW-UHFFFAOYSA-N
chloroacetic anhydride, chloracetic anhydride, monochloroacetic acid anhydride, 2-chloroacetic anhydride, chloroacetic acid anhydride, chloroacetyl anhydride, acetic acid, chloro-, anhydride, unii-8q7txh7r5f, monochloroacetic anhydride, 8q7txh7r5f
10946
(2-chloroacetyl) 2-chloroacetate
ClCC(=O)OC(=O)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 51°C to 59°C
Color Brown
Boiling Point 203°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (ClCH2CO)2O
Quantity 250 g
Beilstein 02, 199
Fieser 01,129
Merck Index 15, 2113
Solubility Information Solubility in water: decomposes. Other solubilities: freely soluble in ether and chloroform,slightly soluble in benzene,practically insoluble in cold ligroin
Formula Weight 170.98
Percent Purity 97%
Physical Form Crystals, Chunks or Low Melting Mass
Chemical Name or Material chloroacetic anhydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'