missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroacetic Acid, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
Specifications
Melting Point | 62°C |
Color | White |
pH | lt; 1 |
Boiling Point | 190°C |
Packaging | Glass Jar |
Quantity | 500 g |
Formula Weight | 94.5g/mol |
Physical Form | Flaky Solid |
Safety and Handling
Product Identifier
- Chloroacetic acid
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H290-May be corrosive to metals.
- H410-Very toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'