Learn More
Chloroacetic acid, 99+%, ACS reagent, ACROS Organics™
Brand: Acros Organics 448110025
Code : 29
Additional Details : CAS Number : 79-11-8 Weight : 3.12500kg Transport : UN number : 1751 Chem class : 6.1 Pack group : II
Packaging | Plastic bottle |
---|---|
Quantity | 2.5kg |
Chemical Identifiers
79-11-8 | |
94.5 | |
chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure | |
CHEBI:27869 | |
C(C(=O)O)Cl |
C2H3ClO2 | |
FOCAUTSVDIKZOP-UHFFFAOYSA-N | |
300 | |
2-chloroacetic acid |
Specifications
Chloroacetic acid | |
1.5800g/mL | |
0.01% max | |
1.58 | |
1.29 Pas (100°C) | |
61.0°C to 63.0°C | |
79-11-8 | |
C2H3ClO2 | |
15,2112 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether | |
C(C(=O)O)Cl | |
94.5 | |
CHEBI:27869 | |
99+% |
0.01% max | |
126°C | |
Plastic bottle | |
0.02% max | |
189.0°C | |
2.5kg | |
99+% | |
ClCH2CO2H | |
chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure | |
FOCAUTSVDIKZOP-UHFFFAOYSA-N | |
2-chloroacetic acid | |
300 | |
94.5 | |
ACS Reagent |
Safety and Handling
- Chloroacetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H311-Toxic in contact with skin.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.