missing translation for 'onlineSavingsMsg'
Learn More

Chloro(triphenylphosphine)gold(I), Light yellow powder, Premion™, 99.99% (Metal basis, Au 39.3% min.), Alfa Aesar™

Brand:  Alfa Aesar 041007.03

Code : NEW

Additional Details : CAS Number : 14243-64-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11319407

  • £193.80 / 1g


Estimated Shipment date
from Supplier 23-01-2018
Add to basket

Specifications

Chemical Identifiers

Chloro(triphenylphosphine)gold(I)
Powder
AuCl[P(C6H5)3]
MFCD00009588
14243-64-2
99.99% (Metal basis, Au 39.3% min.)
494.71
Triphenylphosphinegold(I) chloride

Specifications

Chloro(triphenylphosphine)gold(I)
1g
Odorless
99.99% (Metal basis, Au 39.3% min.)
494.71
Triphenylphosphinegold(I) chloride
Light yellow
14243-64-2
Powder
AuCl[P(C6H5)3]
MFCD00009588
Videos
SDS