missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals

Catalog Number 10478630
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
104-82-5
C8H9Cl
140.61
MFCD00000919
DMHZDOTYAVHSEH-UHFFFAOYSA-N
4-methylbenzyl chloride, p-xylyl chloride, 1-chloromethyl-4-methylbenzene, alpha-chloro-p-xylene, p-methylbenzyl chloride, benzene, 1-chloromethyl-4-methyl, p-chloromethyl toluene, 4-chloromethyl toluene, p-tolylmethyl chloride, p-xylene, alpha-chloro
7722
1-(chloromethyl)-4-methylbenzene
CC1=CC=C(C=C1)CCl
This item is not returnable. View return policy
104-82-5
C8H9Cl
140.61
MFCD00000919
DMHZDOTYAVHSEH-UHFFFAOYSA-N
4-methylbenzyl chloride, p-xylyl chloride, 1-chloromethyl-4-methylbenzene, alpha-chloro-p-xylene, p-methylbenzyl chloride, benzene, 1-chloromethyl-4-methyl, p-chloromethyl toluene, 4-chloromethyl toluene, p-tolylmethyl chloride, p-xylene, alpha-chloro
7722
1-(chloromethyl)-4-methylbenzene
CC1=CC=C(C=C1)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 4°C
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 200°C to 202°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2Cl
Refractive Index 1.5330 to 1.535
Quantity 500 mL
Beilstein 05, 384
Merck Index 15, 10289
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol and ether
Formula Weight 140.61
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material α-Chloro-p-xylene, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a-Chloro-p-xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'