missing translation for 'onlineSavingsMsg'
Learn More

chloro(diethyl)phosphine, 95%, ACROS Organics™

Brand:  Acros Organics 350310050

Code : 29

Additional Details : CAS Number : 686-69-1 Weight : 0.06000kg Transport : UN number : 3129 Chem class : 1a Pack group : II


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10206923

  • £160.20 / 5g


Estimated Shipment
20-02-2018
Add to basket


Specifications

Chemical Identifiers

Chloro(diethyl)phosphine
C4H10ClP
124.55
95%
(CH3CH2)2PCl
MFCD00013621

Specifications

Chloro(diethyl)phosphine
18°C
Clear colorless to light yellow
686-69-1
94% min.
1.4740 to 1.4760
95%
C4H10ClP
124.55
1.0230g/mL
131.0° to 132.0°C
5g
Authentic
Glass bottle
Liquid
95%
(CH3CH2)2PCl
MFCD00013621
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Chloro(diethyl)phosphine

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS