missing translation for 'onlineSavingsMsg'
Learn More

Chloro(diethyl)phosphine, Thermo Scientific Chemicals

Catalog Number 15451167
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
686-69-1
C4H10ClP
124.548
MFCD00013621
INJBDKCHQWVDGT-UHFFFAOYSA-N
chlorodiethylphosphine, diethylchlorophosphine, chloro diethyl phosphine, phosphinous chloride, diethyl, pubchem6480, chloro diethyl phosphane, acmc-1avkn, diethylphosphinous chloride, diethylphosphinous chloride #, chlorodiethyl phosphine
69639
chloro(diethyl)phosphane
CCP(CC)Cl
This item is not returnable. View return policy
686-69-1
C4H10ClP
124.548
MFCD00013621
INJBDKCHQWVDGT-UHFFFAOYSA-N
chlorodiethylphosphine, diethylchlorophosphine, chloro diethyl phosphine, phosphinous chloride, diethyl, pubchem6480, chloro diethyl phosphane, acmc-1avkn, diethylphosphinous chloride, diethylphosphinous chloride #, chlorodiethyl phosphine
69639
chloro(diethyl)phosphane
CCP(CC)Cl

Chloro(diethyl)phosphine is used as reagent. It is used as chemical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.023 g/mL
Boiling Point 131°C to 132°C
Flash Point 18°C (66°F)
Linear Formula (C2H5)2PCl
Refractive Index 1.475
Quantity 1 g
UN Number UN2845
Sensitivity Air and moisture sensitive
Solubility Information Reacts violently.
Formula Weight 124.55
Physical Form Liquid
Chemical Name or Material Chloro(diethyl)phosphine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'