missing translation for 'onlineSavingsMsg'
Learn More

Chloro Bis-(4-fluorophenyl)methane 98%, ACROS Organics™

Brand:  Acros Organics 134960100

Code : 29

Additional Details : CAS Number : 27064-94-4 Weight : 0.06000kg


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10775621

  • £65.20 / 10g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

Chloro bis-(4-fluorophenyl)methane
27064-94-4
98%
238.66
1, 1'-(Chloromethylene)bis(4-fluorobenzene)
98%
≥97.5%
C13H9ClF2
MFCD00044329

Specifications

Chloro bis-(4-fluorophenyl)methane
10g
27064-94-4
Glass bottle
Liquid
98%
238.66
1, 1'-(Chloromethylene)bis(4-fluorobenzene)
Clear colorless to slightly yellow
98%
Authentic
1.5560 to 1.5580 (20°C, 589nm)
≥97.5%
C13H9ClF2
MFCD00044329
Videos
SDS