missing translation for 'onlineSavingsMsg'
Learn More

Chloro bis-(4-fluorophenyl)methane, 98%, Thermo Scientific™

Catalog Number p-85818
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
27064-94-4
C13H9ClF2
238.66
MFCD00044329
FHPNLCLHMNPLEW-UHFFFAOYSA-N
4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
2723773
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F
This item is not returnable. View return policy
27064-94-4
C13H9ClF2
238.66
MFCD00044329
FHPNLCLHMNPLEW-UHFFFAOYSA-N
4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
2723773
1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F
Color Colorless to Yellow
Density 1.2500g/mL
Boiling Point 88°C (0.1 Mbar)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5560 to 1.558
Quantity 10 g
Specific Gravity 1.25
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol,ethanol,2-propanol and,acetone
Formula Weight 238.66
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloro bis-(4-fluorophenyl)methane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Chloro bis-(4-fluorophenyl)methane
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'