missing translation for 'onlineSavingsMsg'
Learn More

chloro(1,5-hexadiene)rhodium(I) dimer, Thermo Scientific™

Catalog Number p-3212499
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
32965-49-4
C12H20Cl2Rh2
441.02
MFCD00064595
This item is not returnable. View return policy
32965-49-4
C12H20Cl2Rh2
441.02
MFCD00064595
Melting Point 118.0°C to 120.0°C
Color Golden
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 mg
Formula Weight 441.02
Physical Form Crystalline Powder
Chemical Name or Material chloro(1, 5-hexadiene)rhodium(I) dimer

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'