missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Chlormezanone, 98+%

Skeletal muscle relaxant

Brand:  Alfa Aesar™ J62934.06

Code : NEW

Additional Details : CAS Number : 80-77-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15465229

  • £32.10 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Chlormezanone
C11H12ClNO3S
MFCD00143951
2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
80-77-3
273.74
14,2106

Specifications

Chlormezanone
114°C
80-77-3
≥98%
273.74
14,2106
White
5g
Powder
C11H12ClNO3S
MFCD00143951
2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one
Videos
SDS