Learn More
Chlormethiazole hydrochloride, Tocris Bioscience™
Potentiates GABAA receptor function
Brand:  RD Systems (Bio-Techne) 88150
Code : Z1
Additional Details : Weight : 0.00050kg
Quantity 50mg
 View more versions of this product
Product Code. 13458103
  • £279.85 / 50mg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Sedative and anticonvulsant which is neuroprotective in a number of animal models. Prevents the degeneration of serotonergic nerve terminals induced by MDMA («Ecstasy«).
Specifications
Chemical Identifiers
6001-74-7
198.105
chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1
5-(2-chloroethyl)-4-methyl-1,3-thiazole;hydrochloride
C6H9Cl2NS
OFXYKSLKNMTBHK-UHFFFAOYSA-N
19035073
CC1=C(SC=N1)CCCl.Cl
Specifications
Chlormethiazole hydrochloride
6001-74-7
chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1
CC1=C(SC=N1)CCCl.Cl
198.105
198.11
50mg
C6H9Cl2NS
OFXYKSLKNMTBHK-UHFFFAOYSA-N
5-(2-chloroethyl)-4-methyl-1,3-thiazole;hydrochloride
19035073
>99%
Videos